missing translation for 'onlineSavingsMsg'
Learn More

Thiamine hydrochloride, Dr. Ehrenstorfer

Product Code. 30320792
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320792 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320792 Supplier Dr. Ehrenstorfer Supplier No. DREC17455000100MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiamine Hydrochloride
CAS 67-03-8
Analyte or Component Names Thiamine Hydrochloride
Shipping Condition +20°C
Molecular Formula C12 H17 N4 O S . Cl . Cl H
Certifications/Compliance ISO 17034
Synonym Thiamine Hydrochloride, Vitamin B1, Thiamine monohydrochloride, 3-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methylthiazolium chloride monohydrochloride, Aneurine hydrochloride, Apate drops, Beatine, Bedome, Begiolan, Benerva, Bequin, Berin, Betabion, Betalin S, Betaxin, Bethiazine, Beuion, Bevitine, Bewon, Biamine, Biuno, Bivatin, Bivita, Clotiamina, Eskapen, Eskaphen, Lixa-Beta, Metabolin, Slowten, THD, Thiadoxine, Thiamin chloride, Thiamin dichloride, Thiamin hydrochloride, Thiaminal, Thiamine chloride, Thiamine chloride hydrochloride, Thiamine dichloride, Thiamine hydrochloride, Thiaminium chloride, Thiaminium chloride hydrochloride, Thiamol, Thiavit, Tiamidon, Tiaminal, Trophite, Vetalin S, Vinothiam, Vitamin B1 hydrochloride, Vitaneuron, 3-(4-Amino-2-methylpyrimidyl-5-methyl)-4-methyl-5,beta-hydroxyethylthiazolium chloride hydrochloride
InChI Formula InChI=1S/C12H17N4OS.2ClH/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;;/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);2*1H/q+1;;/p-1
SMILES Cl.[Cl-].Cc1ncc(C[n+]2csc(CCO)c2C)c(N)n1
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
Molecular Weight (g/mol) 337.27
Quantity 100 mg
Formula Weight 336.0578 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.