missing translation for 'onlineSavingsMsg'
Learn More

Thiamine-13C3 Hydrochloride, TRC

Product Code. 30008095
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008095 10 mg 10mg
30008287 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008095 Supplier Toronto Research Chemicals Supplier No. TRCT34418810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C913C3H18Cl2N4OS
Molecular Weight (g/mol) 340.25
Synonym 3-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-chloridethiazolium-13C3 Hydrochloride, Aneurine-13C3 Hydrochloride, Apate Drops-13C3, Beatine-13C3, Bedome-13C3, Begiolan-13C3, Benerva-13C3, Eskapen-13C3, Eskaphen-13C3, Thiamin-13C3 Dichloride, Vinothiam-13C3, Vitamin B1-13C3 Hydrochloride, Vitaneuron-13C3;
SMILES OCC[13C]1=[13C]([13CH3])[N+](CC2=CN=C(C)N=C2N)=CS1.[Cl-].Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H17N4OS.2ClH/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;;/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);2*1H/q+1;;/p-1/i1+1,8+1,11+1;;
Percent Purity >95%
Chemical Name or Material Thiamine-13C3 Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.