missing translation for 'onlineSavingsMsg'
Learn More

Thiamet G, MedChemExpress

Product Code. 16473316
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473316 25 mg 25mg
16427977 5 mg 5mg
16421467 50 mg 50mg
16415258 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473316 Supplier Medchem Express Supplier No. HY12588
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Thiamet G is a potent and selective inhibitor of O-GlcNAcase (OGA), which acts to remove O-GlcNAc from modified proteins, with Ki of 20 nM for human OGA.

Thiamet G is a potent and selective inhibitor of O-GlcNAcase (OGA), which acts to remove O-GlcNAc from modified proteins, with Ki of 20 nM for human OGA.

Specifications

Chemical Name or Material Thiamet G
Physical Form Powder
CAS 1009816-48-1
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C9H16N2O4S
For Use With (Application) Cancer-Kinase/protease
Solubility Information H2O : ≥ 50 mg/mL (201.37 mM) ∣DMSO : ≥ 45 mg/mL (181.23 mM)
SMILES O[C@H]1[C@H](O)[C@@]2([H])N=C(NCC)S[C@@]2([H])O[C@@H]1CO
Molecular Weight (g/mol) 248.3
Formula Weight 248.3
Percent Purity 98.32%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.