missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THG1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15208-1-AP-20UL
This item is not returnable.
View return policy
Description
Adds a GMP to the 5′-end of tRNA(His) after transcription and RNase P cleavage. This step is essential for proper recognition of the tRNA and for the fidelity of protein synthesis.Specifications
| THG1L | |
| Polyclonal | |
| Unconjugated | |
| THG1L | |
| ICF45, IHG-1, THG1L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 303067, 54974, 66628 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5M965, Q9CY52, Q9NWX6 | |
| THG1L | |
| THG1L Fusion Protein Ag7361 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction