missing translation for 'onlineSavingsMsg'
Learn More

THG1L Antibody (4F11), Novus Biologicals™

Product Code. 18397829 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397829 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397829 Supplier Novus Biologicals Supplier No. H00054974M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

THG1L Monoclonal antibody specifically detects THG1L in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen THG1L
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4F11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH01523
Gene Alias EC 2.7.7, EC 2.7.7.6, FLJ11601, FLJ20546, ICF45EC 2.7.7.-, Interphase cytoplasmic foci protein 45, probable tRNA(His) guanylyltransferase, tRNA-histidine guanylyltransferase, tRNA-histidine guanylyltransferase 1-like (S. cerevisiae)
Host Species Mouse
Immunogen THG1L (AAH01523, 1 a.a. ~ 269 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAKSKFEYVRDFEADDTCLAHCWVVVRLDGRNFHRFAEKHNFAKPNDSRALQLMTKCAQTVMEELEDIVIAYGQSDEYSFVFKRKTNWFKRRASKFMTHVASQFASSYVFYWRDYFEDQPLLYPPGFDGRVVVYPSNQTLKDYLSWRQADCHINNLYNTVFWALIQQSGLTPVQAQGRLQGTLAADKNEILFSEFNINYNNELPMYRKGTVLIWQKVDEVMTKEIKLPTEMEGKKMAVTRTRTKPVPLHCDIIGDAFWKEHPEILDEDS
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 54974
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.