missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Clifton™ Thermostatic Thermostirrer

Product Code. 11479529
Click to view available options
For Use With (Equipment):
Unheated Tanks
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11479529

Brand: Nickel Electro™ NE4T

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Nickel Electro™ Clifton™ Thermostatic Thermostirrer is a durable stainless steel construction, painted surfaces are powder painted with tough anti-bacterial finish with good chemical resistance.

  • Thermostirrer produces a stable temperature environment.
  • Suits a wide range of bath sizes from 8 litre up to 28 litre for large capacity work.
  • Operates in temperature range ambient to 99°C.
  • Quiet running stirrer and good water circulation in the bath.
  • Bold temperature controls, easily seen.
  • Broad range of accessories available to suit wide range of applications.
  • Sturdy and reliable.
  • Built to latest safety standards and supplied with comprehensive operating instructions and Three-year warranty.
  • Before leaving the factory each water bath undergoes following:
    Uniquely identified with its own serial number recorded on our product database, which includes all of its manufacturing inspection data.
  • Electrically tested for safety at final inspection and a test certificate enclosed in rear of the instruction book.
  • To verify the performance of the temperature control system every unit undergoes a factory calibration.
  • Powder painted body providing a durable finish and featuring active anti-bacterial technology.
  • This paint finish actively inhibits bacterial growth improving hygiene requirements as demanded by many clinical and laboratory environments.
  • Hygienic coatings are part of a controlled approach to cleaner facilities.

Specifications

Capacity (Metric) 8 to 28 L
For Use With (Equipment) Unheated Tanks
Voltage 230 V
Frequency 50/60 Hz
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.