missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Chemical Resistant Pan

Product Code. 15339433 Shop All Cole-Parmer Products
Change view
Click to view available options
Dimensions (L x W x H):
19.5 x 19.5 x 1.25 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
15339433 19.5 x 19.5 x 1.25 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15339433 Supplier Cole-Parmer™ Supplier No. 0671588
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Resistant Pan is resistant to mild acids, alkaline solutions and many reagents.

  • Withstand temperatures from -40°F to +250°F (-40°C to +121°C).
  • Contain spills with straight edge pans.
  • Constructed of Thermoset polyester resin, these white trays are ideal for carrying, storing, and cleaning of lab product.
  • Trays withstands temperatures from 40°C to 121° (-40°F to +250°F).
  • Pans with straight edges are ideal for spill containment.

Specifications

Dimensions (L x W x H) 19.5 x 19.5 x 1.25 in.
Color White
Height (Metric) 3.2 cm
Length (Metric) 49.5 cm
Material Plastic
Type Laboratory Tray
Width (Metric) 49.5 cm
For Use With Ideal for carrying, storing, and cleaning of lab products
Length (English) 19.5 in.
No. per Pack 1 Ea.
Height (English) 1.25 in.
Shape Square
Width (English) 19.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.