missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ThermoPrime Taq DNA Polymerase, with 10X buffer and separate vial of 25 mM MgCl2

Product Code. 11727227
Change view
Click to view available options
Quantity:
2500 units
Unit Size:
2500 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11727227 2500 units 2500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11727227 Supplier Thermo Scientific™ Supplier No. AB0301B
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific ThermoPrime Taq Taq Polymerase is a 94 kDa recombinant thermostable DNA polymerase obtained by high level expression of the Taq DNA polymerase gene in E. coli.

Thermo Scientific ThermoPrimeTaq Polymerase is a 94-kDa recombinant thermostable DNA polymerase obtained by high level expression of the Taq DNA Polymerase gene in E. coli. A proprietary buffer component protects ThermoPrime DNA Polymerase during high temperature steps, ensuring optimal performance throughout the reaction, leading to increased PCR yields. Convenient master mix format is also available for easier reaction set-up.

Features

  • Enhanced enzyme stability for increased yield
  • Advanced buffering system minimizes PCR optimization steps

Applications

  • Amplification of fragments up to 5 kb
  • Routine PCR
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage • ThermoPrime Taq DNA Polymerase (5 U/μL), 10 x 50 μL
• 10X Reaction Buffer IV, 10 x 1.5 mL
• 25 mM MgCl2, 10 x 1.5 mL

Store at -20°C.
GC-Rich PCR Performance Low
Polymerase ThermoPrime Taq DNA Polymerase
Reaction Speed Standard
Product Type DNA Polymerase
Quantity 2500 units
Shipping Condition Wet or Dry Ice
For Use With (Application) Standard PCR
Fidelity (vs. Taq) 1X
Hot Start No
No. of Reactions 4000 Reactions
Overhang 3'-A
Reaction Format Separate Components
Starting Material DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.