missing translation for 'onlineSavingsMsg'
Learn More

VEE GEE Thermometer, Encapsulated in FEP

Product Code. 17871254 Shop All Thermo Scientific CO₂ Incubators Products
Change view
Click to view available options
Temperature Range (English):
-30 to +120°F
-31 to +122°F
-4 to +230°F
-4 to +302°F
0 to 300°F
30 to 220°F
Subdivision:
1°C (1.8°F)
1.1°C (2°F)
Immersion Length:
30.4 cm (12 in.)
30.48 cm (12 in.)
39 cm (15.4 in.)
To immersion line
Immersion Type:
Partial Immersion
Total Immersion
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (English) Subdivision Immersion Length Immersion Type unitSize
17871254 -30 to +120°F 1.1°C (2°F) 30.48 cm (12 in.) Total Immersion Each
17851194 -4 to +230°F 1.1°C (2°F) 30.48 cm (12 in.) Total Immersion Each
17811244 0 to 300°F 1.1°C (2°F) 30.48 cm (12 in.) Total Immersion Each
17831234 30 to 220°F 1.1°C (2°F) 39 cm (15.4 in.) Total Immersion Each
17881244 -31 to +122°F 1°C (1.8°F) 30.48 cm (12 in.) Total Immersion Each
17841244 -4 to +230°F 1°C (1.8°F) 30.4 cm (12 in.) Total Immersion Each
17881204 -4 to +302°F 1°C (1.8°F) 30.48 cm (12 in.) Total Immersion Each
17801234 -30 to +120°F 1.1°C (2°F) To immersion line Partial Immersion Each
17831204 -4 to +230°F 1.1°C (2°F) To immersion line Partial Immersion Each
17811214 0 to 300°F 1.1°C (2°F) To immersion line Partial Immersion Each
17841214 30 to 220°F 1.1°C (2°F) To immersion line Partial Immersion Each
17871234 -31 to +122°F 1°C (1.8°F) To immersion line Partial Immersion Each
17851214 -4 to +230°F 1°C (1.8°F) To immersion line Partial Immersion Each
17881214 -4 to +302°F 1°C (1.8°F) To immersion line Partial Immersion Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 17871254 Supplier VEE GEE Supplier No. 80701E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The encapsulated glass thermometer is designed to safeguard against broken glass and liquid. It contains a biodegradable Safety Blue fill, offering mercury-like properties minus the associated risks of contamination and hazards.

  • FEP encapsulation- in the event of breakage the FEP coating contains the broken glass and liquid, preventing injury or contamination
  • Feature Safety Blue fill against a white background for easy reading
  • Accuracy is equal to one subdivision
  • Immersion: Total or Partial

Specifications

Bottle Fill Safety Blue
For Use With (Application) Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications
Immersion Type Total Immersion
Length (Metric) 30.48 cm
Material Glass
Probe Type Immersion/General Purpose (Tapered)
Thermometer Fill Safety Blue
Weight (Metric) 5 g
Product Type Serialized Glass Thermometer
Max. Temperature (English) 120°F
Max. Temperature (Metric) 48.9°C
Min. Temperature (English) -30°F
Min. Temperature (Metric) -34.4°C
Temperature Range (English) -30 to +120°F
Temperature Range (Metric) -34.4 to +48.9°C
Accuracy ±1 subdivision
Subdivision 1.1°C (2°F)
Immersion Length 30.48 cm (12 in.)
Width (Metric) 1.52 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.