Learn More
Dostmann Electronic™ Digital Thermometer
Product Code. 11814372
Change view
Click to view available options
Calibrated:
Yes
Unit Size:
Each
Product Code. Calibrated Unit Size
11814372 Yes Each
1 options
Product Code. 11814372 Supplier Dostmann Electronic™ Supplier No. 50000700
  • ONLINE EXCLUSIVE PRICE
    £473.02
    £219.28 / Pack of 1
    Save £253.74
    53% Off
View all promotions
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
With temperature measurements over a range of -200°C to +850°C, this digital thermometer is ideal for use in many laboratory, scientific, and other research environments.
  • The all-round talent for temperature measurements with Pt100 sensor over a range of -200°C to +850°C
  • The thermocouple (type J, K, L, N, R, S, T) to +1760°C with a resolution of 0,1°C over the full measuring range
  • Ideal for quality assurance service and production
  • Integrated calibration function for simple compensation of sensor tolerances
  • Physical 1-point, 2-point or 3-point calibration function
Specifications
Calibrated Yes