Learn More
Thermo Scientific™ Thermolyne™ Premium Large Muffle Furnace, digital programmable control with 4 programs, 16 segments each, D1, 240V
Ideal for industrial applications, Thermolyne Premium Large Muffle Furnace features a robust design and a choice of four temperature controller models.
Brand:  Thermo Scientific™ F6030CM-33
Product Code. 12441892
  • ONLINE EXCLUSIVE PRICE
    £5115.00
    £4773.50 / Each
    Save £341.50
    6% Off
Only null left
Estimated Shipment: 25-06-2026
Add to basket
Description
Description
Ideal for industrial applications, Thermo Scientific™ Thermolyne Premium Large Muffle Furnace features a robust design and a choice of four temperature controller models. This furnace provides a spacious 14L (0.5 cu. ft.) capacity and reaches a maximum of 1200°C, making it ideal for ashing organic and inorganic samples and conducting gravimetric analysis.
  • Four heating elements are located on the chamber top, bottom and sides for enhanced temperature uniformity
  • Built-in vent port removes undesirable contaminants and moisture to extend the life of the element and unit
  • Rear of the chamber incorporates a 0.95cm (0.38in.) dia. port for monitoring chamber temperatures with independent measuring devices
  • Optional stainlesssteel shelf doubles load capacity (maximum temperature of 900°C)
  • Door safety switch stops power to heating elements when door opens
  • Thermocouple break protection cuts power to the heating elements, preventing a thermocouple failure runaway condition
  • Furnaces with B1, C1, and D1 control also use a mechanical overtemperature protection relay
Choice of Temperature Controllers
  • Digital single setpoint control (A1): Single display shows actual temperature or setpoint
  • Digital single setpoint control with single ramp to setpoint and a dwell (B1): Single display shows actual temperature or setpoint
  • Digital programmable control with one stored program of eight segments (C1)
  • Digital programmable control with four stored programs, 16 segments per program (D1): Controller includes RS-232 communications
Includes:
Models F6010 and F6018 include a cord and plug set.
Requires:
All except Models F6010 and F6018 require a conduit connection.
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products