missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HUBER Thermofluid Ethylenglykol
For use with chillers
£450.50 - £1653.60
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10488201
|
HUBER
6170 |
10 L |
£450.50
10 L |
Estimated Shipment: 20-11-2024
Log in to see stock. |
|||||
11597354
|
HUBER
6171 |
50 L |
£1653.60
50 L |
Estimated Shipment: 20-11-2024
Log in to see stock. |
|||||
Description
Thermofluid - Ethylenglykol, for ChillersSpecifications
Chillers | |
Thermofluid Ethylenglykol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Monoethylenglykol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P314-Get medical advice/attention if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501-Dispose of contents/container to .