missing translation for 'onlineSavingsMsg'
Learn More

HUBER Thermofluid DW-Therm HT

Product Code. 10467642
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
Product Code. Quantity unitSize
10467642 5 L 5 L
1 options
This item is not returnable. View return policy

Product Code. 10467642

Brand: HUBER 6672

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Valid only for Unistats.

  • Thermofluid DW-Therm HT P20.340.32
  • Unistat-thermofluid for closed systems
  • Min. temp: 20°C
  • Max. temp.: (170)340°C
  • Viscosity at 25°C: 32 mm2/sec

Specifications

Color Orange
Temperature Range 20°C to 340°C
Type Thermofluid DW-Therm HT
Quantity 5 L
Temperature (Metric) Maximum 340°C
Viscosity 32 mm2/sec.
compliance-icons
Product Identifier
  • DW-therm HT (heat transfer fluid)
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P391-Collect spillage.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : 1,2,3,4-tetrahydro(1-phenylethyl)-napthalin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.