Learn More
Thermo Scientific™ Thermo-Start™ Taq DNA Polymerase PCR Kts and Master Mxes
Hot-start modified for PCR applications requiring maximum specificity and sensitivity to detect rare or complex targets and allow for room-temperature reaction setup.
Brand: Thermo Scientific™ AB-1057/B
Code : A0
Additional Details : CAS Number : 7732-18-5 Weight : 0.50000kg
Description
Thermo Scientific™ Thermo-Start mixes are hot-start modified for PCR applications requiring maximum specificity and sensitivity, to detect rare or complex targets; also allows for room-temperature reaction setup.
Thermo-Start Taq DNA Polymerase is a hot-start enzyme combined with an optimized buffer system that gives superior specificity and sensitivity. The chemically modified Thermo-Start Taq remains completely inactive until it is switched on by a high temperature incubation step at the start of the PCR reaction. This prevents nonspecific amplification during reaction setup, eliminating smearing and the formation of nontarget bands.- Hot-Start modification ensures stringent specificity
- Safe for room temperature reaction setup
- Enhanced enzyme stability for increased yield and sensitivity
- Advanced buffering system minimizes PCR optimization steps
- High Performance Buffer formulation for GC-rich and other complex templates
Format Options:
Enzyme Kits are the most basic format. Kits include the enzyme, PCR buffer, and 25mM MgCl2 solution, each in separate vials. dNTPs must be purchased separately
Complete PCR Kits include all the components needed to perform PCR, including an HPLC-purified dNTP mix. Complete PCR kits are a convenient option that still allows for MgCl2 optimization
Master Mix formats consist of a single master buffer containing all the components needed for PCR. Master Mixes are the most convenient format as they significantly reduce assay set-up steps and minimize the risk of sample contamination
Premixed MgCl2 Kits have MgCl2 premixed into the PCR buffer and will be at 1.5mM concentration in the final reaction
High Performance Buffer Kits are specially formulated for GC-rich or other complex targets with a high degree of secondary structure
Compatible with:
Thermo-Start Taq Reddymix Kits and Master Mixes
Recommended for:
Amplifying fragments up to 5kb; Hot-Start PCR; RT-PCR; Sequencing; Multiplexing; Genotyping; Library Construction; Cloning; Pathogen Detection; Single Cell PCR
Specifications
Store at -20°C for long-term storage. Stable at 4°C for up to four weeks. Avoid freeze-thaw cycles. | |
4000 x 25µL Rxns | |
Thermo-Start Taq Reddymix Kits and Master Mixes |
Thermo-Start DNA Polymerase Enzyme Kit w/High Performance Buffer (for GC-rich) | |
Cloning | |
5U/µL |
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.