missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Thermo Sequenase Cycle Sequencing Kit

Product Code. 15819916
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15819916 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819916 Supplier Applied Biosystems™ Supplier No. 785001KT

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Thermo Sequenase with thermostable Pyrophosphatase, Control DNA pUC18, Control Primer (-40 Forward, 23-mer), Cycle Mix 7-deaza-dGTP, Cycle Mix dATP, Cycle Mix dTTP, Cycle Mix dCTP, ddGTP Termination Mix (7-deaza-dGTP), ddATP Termination Mix (7-deaza-dGTP), ddTTP Termination Mix (7-deaza-dGTP), ddCTP Termination Mix (7-deaza-dGTP), Reaction Buffer (concentrate), Stop Solution, Protocol Book

The Thermo Sequenase Cycle Sequencing Kit allows for equally efficient incorporation of both ddNTPs and dNTPs in cycle sequencing reactions resulting in very uniform band intensities which can be read accurately.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Kit Components:
Thermo Sequenase with thermostable Pyrophosphatase
Control DNA pUC18
Control Primer (-40 Forward, 23-mer)

Cycle Mix 7-deaza-dGTP
Cycle Mix dATP
Cycle Mix dTTP
Cycle Mix dCTP
ddGTP Termination Mix (7-deaza-dGTP)
ddATP Termination Mix (7-deaza-dGTP)
ddTTP Termination Mix (7-deaza-dGTP)
ddCTP Termination Mix (7-deaza-dGTP)

Reaction Buffer (concentrate)
Stop Solution
Protocol Book

This kit and all the enclosed reagents should be stored frozen at -20°C (NOT in a frost-free freezer). Keep all reagents on ice when removed from storage for use.

Description Thermo Sequenase Cycle Sequencing Kit, –20°C Stored
Format Kit
For Use With (Application) DNA Sequencing, Sanger Sequencing
Includes Thermo Sequenase with thermostable Pyrophosphatase, Control DNA pUC18, Control Primer (-40 Forward, 23-mer), Cycle Mix 7-deaza-dGTP, Cycle Mix dATP, Cycle Mix dTTP, Cycle Mix dCTP, ddGTP Termination Mix (7-deaza-dGTP), ddATP Termination Mix (7-deaza-dGTP), ddTTP Termination Mix (7-deaza-dGTP), ddCTP Termination Mix (7-deaza-dGTP), Reaction Buffer (concentrate), Stop Solution, Protocol Book
Product Type Thermo Sequenase Cycle Sequencing Kit
Quantity 1 Kit

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.