Learn More
Thermo Scientific™ Thermo Scientific™ Watch Glass
Product Code. 17145321
Change view
Click to view available options
Material:
Glass
Unit Size:
Pack of 400
Product Code. Material Unit Size
17145321 Glass Pack of 400
1 options
Product Code. 17145321 Supplier Thermo Scientific™ Supplier No. WATCH GLASS
  • ONLINE EXCLUSIVE PRICE
    £233.00
    £225.18 / Pack of 400
    Save £7.82
    3% Off
Estimated Shipment: 15-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Thermo Scientific™ Watch Glass is used as a beaker cover and used in weighing, crystallization, and drying in the lab.
Thermo Scientific™ Watch Glass is used as a beaker cover as well as used in weighing, crystallization, and drying in the lab.
  • WATCH GLASS 2 PER SET
  • 200 SETS /CS
TRUSTED_SUSTAINABILITY
Specifications
Description WATCH GLASS 2 PER SET, 200 SETS /CS
Material Glass
Type Watch Glass
For Use With (Application) Used as Beaker Covers