missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Ultrafast UFC-1 Phase GC Columns
Short, narrow-bore columns for use with the Thermo Scientific TRACE GC Ultra Chromatograph
Brand: Thermo Scientific™ UFMC00502010006
Code : A0
Additional Details : Weight : 1.00000kg
Description
- Reduces analytical run time for increased throughput
- Long column lifetime
Specifications
0.4 μm | |
Ultrafast | |
10 m |