Learn More
Thermo Scientific™ TX-100/100S Clinical Swinging Bucket Rotor Adapters
Shop All Thermo Scientific ProductsDescription
Adapters for TX-100S Clinical Rotor and TX-100 Clinical Rotor
Specifications
Specifications
| Type | Microtube Adapter |
| Tube Type | Microtube |
| Capacity | 1.5/2 mL Microtube |
| For Use With (Equipment) | TX-100 and TX-100S clinical swing bucket rotor |
| Tube Size | 1.5/2 mL |