missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ABgene™ EasyStrip PCR Tubes
Click to view available options
Color:
Clear
White
Closure Type:
Domed Cap
Ultra-clear Flat Cap
Unit Size:
Pack of 250
Description
For QPCR, EasyStrip Snap Tubes feature Ultra Clear Caps tethered to either white or natural tubes. White wells have been proven to increase assay sensitivity and reproducibility. The attached Ultra Clear Cap is an optically clear window into the reaction effectively allowing fluorescent signal to pass through unaffected, facilitating the benefits of white plastic.
In standard PCR, domed caps are used to allow for the best seal when using thermal cyclers with a screw down lid. The domed capped tubes are available in natural. With attached caps, pre- and post- aliquoting steps are much easier to perform. They allow individual access to each tube without compromising any steps in your workflow.
In standard PCR, domed caps are used to allow for the best seal when using thermal cyclers with a screw down lid. The domed capped tubes are available in natural. With attached caps, pre- and post- aliquoting steps are much easier to perform. They allow individual access to each tube without compromising any steps in your workflow.
- 8-strip PCR tubes with individually attached caps
- Each cap can be opened and closed separately, preventing cross-contamination
- The only strip tubes available that combine optimal conditions for QPCR detectiondash (em)white tubes and ultra clear attached caps
- Individually lettered caps help to orient the tubes and aid in verifying sample identity
- Tubes feature uniform, thin walls and are certified free from RNase, DNase, endotoxins and contaminating DNAbullets1 (end)
Specifications
Specifications
| Color | White |
| Quantity | 250 Strips |
| For Use With (Equipment) | 0.2mL Cycler Blocks |
| Shape | Flat |
| Closure Type | Ultra-clear Flat Cap |
| For Use With (Application) | QPCR |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction