missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ABgene™ EasyStrip PCR Tubes

Product Code. 10656154
Click to view available options
Color:
Clear
White
Closure Type:
Domed Cap
Ultra-clear Flat Cap
Unit Size:
Pack of 250
This item is not returnable. View return policy

Product Code. 10656154

Brand: Thermo Scientific™ AB1504

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Each tube in the strip has an individually sealable cap to reduce the chance of sample contamination. PCR and QPCR setup and analysis are made much simpler with the unique design of these new tubes.

For QPCR, EasyStrip Snap Tubes feature Ultra Clear Caps tethered to either white or natural tubes. White wells have been proven to increase assay sensitivity and reproducibility. The attached Ultra Clear Cap is an optically clear window into the reaction effectively allowing fluorescent signal to pass through unaffected, facilitating the benefits of white plastic.

In standard PCR, domed caps are used to allow for the best seal when using thermal cyclers with a screw down lid. The domed capped tubes are available in natural. With attached caps, pre- and post- aliquoting steps are much easier to perform. They allow individual access to each tube without compromising any steps in your workflow.
  • 8-strip PCR tubes with individually attached caps
  • Each cap can be opened and closed separately, preventing cross-contamination
  • The only strip tubes available that combine optimal conditions for QPCR detectiondash (em)white tubes and ultra clear attached caps
  • Individually lettered caps help to orient the tubes and aid in verifying sample identity
  • Tubes feature uniform, thin walls and are certified free from RNase, DNase, endotoxins and contaminating DNAbullets1 (end)
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Quantity 250 Strips
For Use With (Equipment) 0.2mL Cycler Blocks
Shape Domed
Closure Type Domed Cap
For Use With (Application) PCR
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.