Learn More
Epredia™ TissueLoc™ HistoScreen™ Cassettes
The ultimate cassette for specimen security
Brand:  Epredia™ C-1000-RD
Code : A0
Additional Details : Weight : 1.00000kg
Product Code. 12006829
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • For handling small biopsies and fragile specimens
  • Save time, offer the greatest specimen security, and provide ease of use
  • Unique design provides an inner chamber lined with a “see through” mesh for easy identification of specimens
  • Eliminates the need for messy sponges or lens paper
  • Cassettes will not pop open during processing, ensuring your tissues are safely processed