Learn More
Thermo Scientific™ SWAP-IT™ GC-MS Interface System Accessories
Product Code. 10249504 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Ferrule
SWAP-Tight Ferrule 0.1 to 0.25 mm
SWAP-Tight Ferrule 0.32 mm
SWAP-Tight Ferrule, Blank
SWAP-Tight Nut
SWAP-Tight Nut and Adaptor
Transfer Line Nut
Unit Size:
Each
Pack of 10
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10249504 SWAP-Tight Nut and Adaptor Each
10437704 SWAP-Tight Ferrule, Blank Pack of 10
10331055 Transfer Line Nut Each
10351555 SWAP-Tight Nut Each
10117414 Transfer Line Nut Each
10085644 Ferrule Pack of 10
10279264 SWAP-Tight Ferrule 0.1 to 0.25 mm Pack of 10
10249794 SWAP-Tight Ferrule 0.32 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10249504 Supplier Thermo Scientific™ Supplier No. 290SG003
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Find the right nut and ferrule to use with the powerful Thermo Scientific™ SWAP-IT™ GC-MS Interface system.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type SWAP-Tight Nut and Adaptor
Description for S/SL Injector
Quantity 1/Ea.
For Use With (Equipment) S/SL Injector, SWAP-IT GC-MS Interface System for Agilent Instruments
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.