Description
The Thermo Scientific SWAP-IT GCMS Interface provides a powerful tool for switching capillary columns without the need to vent the MS.
- Specially deactivated , robust stainless steel transfer capillary provides an interface between the analytical column and the MS detector
- Small ID of transfer capillary ensures exposure to air and water is reduced to a minimum during disconnection of the analytical column
- The system works without the requirement for additional He-purge/make-up gas flow
- Finger-tight connections make exchanging the analytical column quick and easy without the need for tools
- Includes finger-tight connections for S/SL injector to provide complete finger-tight solution
Specifications
Specifications
| Type | Ferrule |
| Description | Graphite/Vespel Ferrule; For Thermo Scientific Transfer Line |
| Quantity | 10/Pk |
| For Use With (Equipment) | DSQ, DSQ II, PolarisQ, ITQ |