missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Heracell™ Support Frame with Four Casters and Three Drawers

Product Code. 10381092 Shop All Thermo Scientific Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
10381092 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10381092 Supplier Thermo Scientific™ Supplier No. 50056459
  • ONLINE EXCLUSIVE PRICE
    £1870.00
    £1720.49 / Pack of 1
    Save £149.51
    7% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Use With: Heracell 150i CO2 Incubator

  • Provide protection against floor contamination
  • Choice of three heights: 185mm (7.3 in.), 200mm (7.9 in.), or 780mm (30.7 in.)
  • Support frames can also be caster-mounted for easy maneuverability

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Heracell 150i CO2 incubator
Lighting LED
Type Support Frame
Includes Four casters
Height (English) 30.7 in.
Height (Metric) 78 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.