missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Thermo Scientific Sintered Glass LVI Liner
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 09215501
Code : 13
Additional Details : Weight : 0.10000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Thermo Scientific instruments
Specifications
Large Volume Injection (LVI) Liner | |
120 mm | |
2.75 mm | |
Sintered Glass |
1 Ea. | |
0.78, 1.8 mm | |
Thermo Scientific GC Instruments |