Learn More
Thermo Scientific™ Renvo™ SARS-CoV-2 Control Kit
Product Code. 17609995 Shop All Thermo Scientific Products
Product Code. 17609995 Supplier Thermo Scientific™ Supplier No. RENCOV41001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Thermo Scientific™ Renvo™ SARS-CoV-2 Control Kit provides positive and negative control swabs for use with the Thermo Scientific™ Renvo™ SARS-CoV-2 Test. When used for quality control in the test, they are useful as an aid in verifying test and operator performance.
This product has been discontinued and is no longer available for purchase.
The Thermo Scientific™ Renvo™ SARS-CoV-2 Control Kit provides positive and negative control swabs for use with the Thermo Scientific™ Renvo™ SARS-CoV-2 Test. When used for quality control in the test, they are useful as an aid in verifying test and operator performance. Intended use:
For environmental surveillance use only and not for diagnostic purposes.
Contents:
  • SARS-CoV-2 high positive control swab (3): DNA-based synthetic oligo dried onto a swab above the limit of detection of the test.
  • SARS-CoV-2 low positive control swab (3):DNA-based synthetic oligo dried onto a swab above the limit of detection of the test.
  • SARS-CoV-2 negative control swab (3): Buffer solution dried onto a swab.
We recommend running a SARS-CoV-2 positive and negative control:
  • Once for each new lot or shipment of kits received
  • Once for each new operator
TRUSTED_SUSTAINABILITY
Specifications
Description  Provides positive and negative control swabs for use with the Thermo Scientific™ Renvo™ SARS-CoV-2 Test
Includes
  • Three high positive control swabs
  • Three low positive control swabs
  • Three negative control swabs