Learn More
Thermo Scientific™ Remel™ Parabacteroides distasonis ATCC™ 8503, Culti-Loops™
Product Code. 12988858
Click to view available options
Species:
Parabacteroides distasonis
Unit Size:
Pack of 5
Product Code. 12988858
Brand: Thermo Scientific™ R4601226
Perform testing of media, stains, reagents, and identification kits, and evaluate bacteriological procedures with Thermo Scientific ™ Remel™ Parabacteroides distasonis ATCC™ 8503, Culti-Loops™.
The loop may be dissolved in liquid medium, Culti-Loop Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents, and identification kits, and for the evaluation of bacteriological procedures. MycoBacti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms.
Order Info
Culti-Loop products are individually sealed in a foil pouch and packaged five loops per package. Each Mycobacti-Loop is individually sealed in a foil pouch and packaged one loop per package. BactiDisk products are packaged 10 disks per vial.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 8503™*
Product Type Culti-Loops Quality Control Organism
Species Parabacteroides distasonis
Culti-Loops are CE marked.