Learn More
Thermo Scientific™ Remel™ Brevibacillus agri ATCC™ 51663, Culti-Loops™
Product Code. 12945833
Change view
Click to view available options
Species:
Brevibacillus agri
Unit Size:
Pack of 5
Product Code. Species Unit Size
12945833 Brevibacillus agri Pack of 5
1 options
If you would like to order, a signed ATCC agreement is required.
Product Code. 12945833 Supplier Thermo Scientific™ Supplier No. R4607217
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
If you would like to order, a signed ATCC agreement is required.
The loop may be dissolved in liquid medium, Culti-Loop Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents, and identification kits, and for the evaluation of bacteriological procedures. MycoBacti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms.
Order Info
Culti-Loop products are individually sealed in a foil pouch and packaged five loops per package. Each Mycobacti-Loop is individually sealed in a foil pouch and packaged one loop per package. BactiDisk products are packaged 10 disks per vial.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 51663™*
Product Type Culti-Loop
Strain Designations NRS 1219 [13, DSMZ 6348, IFO 15538, JCM 9067, LMG 15103, Vitek #202316]
Species Brevibacillus agri
Culti-Loops are CE marked.