Learn More
Thermo Scientific™ QCC (Quick Column Change) for Agilent Instruments
This Quick Column Change accessory is compatible with Agilent Instrumentation
Brand: Thermo Scientific™ 60180-872
Code : A0
Additional Details : Weight : 0.20000kg
Description
QCC (Quick Column Change)-Leak Free Column Installation
- Tool-free installation of analytical GC column
- Graphite ferrules
Complete QCC Kits Includes:
- Injector Adaptor
- Detector Adaptor
- Tight Nut
- Ferrules (1 each 0.25, 0.32,0.53)
- Ceramic Column Cutter
Specifications
S/SL Injector Adapter | |
S/SL Injector Adaptor |