missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human VEGFR1 Recombinant Protein

Product Code. 11549061
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11549061 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11549061 Supplier Invitrogen™ Supplier No. RP75749

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

RP-75749 contains purified recombinant human VeGF Receptor-1 protein with an N-terminal hexahistidine tag. RP-75749 may be used as a positive control in many immunological assays.

FLT1 (VEGFR1) is a receptor tyrosine kinase which plays a critical role in angiogenesis. Active angiogenesis has been found to play important role in many pathological and physiological processes, such as tumor progression, diabetic retinopathy, rheumatoid arthritis and wound healing. May play an essential role as a negative regulator of embryonic angiogenesis by inhibiting excessive proliferation of endothelial cells. Can promote endothelial cell proliferation, survival and angiogenesis in adulthood. Its function in promoting cell proliferation seems to be cell-type specific. Promotes PGF-mediated proliferation of endothelial cells, proliferation of some types of cancer cells, but does not promote proliferation of normal fibroblasts (in vitro). Has very high affinity for VEGFA and relatively low protein kinase activity; may function as a negative regulator of VEGFA signaling by limiting the amount of free VEGFA and preventing its binding to KDR.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P17948
Concentration 0.75 mg/mL
For Use With (Application) Control, Western Blot
Formulation 25 mM sodium acetate with 50% glycerol and no preservative; pH 4.8
Gene ID (Entrez) 2321
Name Human VEGFR1
pH Range 4.8
Quantity 100 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias AI323757; embryonic receptor kinase 2; Emrk2; FLT; FLT1; FLT-1; fms related tyrosine kinase 1; FMS-like tyrosine kinase 1; FMS-related tyrosine kinase 1; Fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); FRT; i21; sFlt1; soluble VEGFR 1; sVEGFR 1; Tyrosine-protein kinase FRT; tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; vascular endothelial growth factor receptor-1; vascular permeability factor receptor; VEGF R1; VEGFR1; VEGFR-1
Common Name VEGF Receptor 1
Gene Symbol FLT1
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Asp31-Ile328 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
compliance-icons
Product Identifier
  • Human VEGFR1 Recombinant Protein
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Glycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.