missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TIMP1 Recombinant Protein
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
This product contains a purified 184 amino acid fragment (aa24-207) corresponding to the mature TIMP-1 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
TIMP1 (metalloproteinase inhibitor 1) is a metalloproteinase inhibitor that functions by forms one-to-one complexes with target metallproteinases, such as collagenases, and irreversible inactivates them by binding to their catalytic zinc cofactor. TIMP1 acts on MMP1, MMP2, MMP3, MMP7, MMP8, MMP9, MMP10, MMP11, MMP12, MMP14, and MMP16. It also functions as a growth factor that regulates cell differenation, migration, and cell death. TIMP1 activates cellular signaling cascades via CD63 and ITGB1.
Specifications
Specifications
| Accession Number | P01033 |
| Concentration | 0.29 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 25 mM sodium acetate with 50% glycerol and no preservative; pH 4.8 |
| Gene ID (Entrez) | 7076 |
| Molecular Weight (g/mol) | 25.21 kDa |
| Name | Human TIMP1 |
| pH Range | 4.8 |
| Quantity | 100 μg |
| Source | E. coli |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction