missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RBP4 Recombinant Protein
Description
This product contains purified recombinant human Retinol Binding Protein protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
This protein belongs to the lipocalin family and is the specific carrier for retinol in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein post translationally and results in defective delivery and supply to the epidermal cells.
Specifications
Specifications
| Accession Number | P02753 |
| Concentration | 1.0 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 20 mM tris HCl with 50% glycerol and no preservative; pH 8 |
| Gene ID (Entrez) | 5950 |
| Name | Human RBP4 |
| Quantity | 1 mg |
| Source | E. coli |
| Immunogen | N/A |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?