missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IgM Isotype Control

Product Code. 10076623
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10076623 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10076623 Supplier Invitrogen™ Supplier No. 31146

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Polyclonal Isotype Control

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Purity: The precipitin lines against goat anti-human whole serum are the same as those against goat anti-human IgM, Fc5-mu fragment specific, when assayed by immunoelectrophoresis at an antigen concentration equivalent to 20 mg/mL. No reaction was observed against goat anti-human IgG, Fc fragment specific, goat anti-human IgA, alpha chain specific, or goat anti-human alpha-2-macroglobulin. This antibody is recommended for use in blocking assays, immunohistochemical, immunocytochemical and immunofluorescent applications. Storage: Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of Origin: USA.

IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgM
Applications Control, Inhibition Assays, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 4.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.6
Gene Alias AGM1; IGHM; immunoglobulin heavy constant mu; MU; VH
Host Species Human
Purification Method Purified
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 4°C
Product Type Isotype Control
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.