missing translation for 'onlineSavingsMsg'
Learn More

Phalloidin Control, DyLight 550, Invitrogen™

Control

Brand:  Invitrogen 21835

Product Code. 13414309

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Phalloidin Control, DyLight 550 conjugate, 21835, from Invitrogen., 21835, from Invitrogen. Species Reactivity: Many; Applications: Immunocytochemistry, Immunofluorescence, Immunohistochemistry.

Phalloidin is a bicyclic peptide that belongs to a family of toxins isolated from the deadly Amanita phalloides “death cap” mushroom and is commonly used as a counterstain (similar to DAPI or Hoechst) in cell biology and histology imaging applications to selectively label F-actin in fixed cells, permeabilized cells, and cell-free experiments. Labeled phalloidin conjugates have similar affinity for both large and small filaments and bind in a stoichiometric ratio of about one phallotoxin per actin subunit in both muscle and non-muscle cells. Phalloidins reportedly do not bind to monomeric G-actin, unlike some antibodies against actin. The dynamics of the actin polymerization in cells are important for a variety of cellular processes from cell motility to cell shape, from muscular contraction to cytokinesis, and more.
Cross Reactivity: Many Species
Formulation: 300 units purified control antibody (300 units/ml) in methanol and no preservative
Synonyms: DY550-Phalloidin, F-actin toxin, Amanita phalloides
Format: 300 units of lyophilized DyLight 550-Phalloidin (300 units/ml) in methanol. Prepare stock solution by adding 1000µl of pure methanol to the vial and gently mix. 21835 has been successfully used in immunofluorescence and immunohistochemistry. DyLight 550-Phalloidin has an excitation/emission of 557/573nm and molecular weight of 1452.68 g/mole. DyLight 550-Phalloidin Stock Solution is prepared in methanol, and is also soluble in DMF, or DMSO. The unused Stock Solution should be promptly stored at -20°C in a foil pouch with desiccant, and protected from light. The stock solutions are stable for at least one year. Staining cells with some of the DyLight Phalloidin conjugates may require the use of a higher concentration of the phalloidin conjugate; therefore to minimize the amount of methanol added to the cells, the vial contents could be dissolved in 0.5mL methanol to yield a final concentration of 600units/mL. Working solution of 1unit/mL (per 96-well plate) can be prepared by diluting 20µl of the Stock Solution in 5.98mL of PBS and mixing well. Typical staining procedure adds 50µl of Working Solution (i.e. 1 to 5 unit/ml) to a each well. Incubate cells in the dark for 30 minutes at room temperature (optimal staining times varies from 10 minutes to 3 hours depending on cell type). Aspirate and wash cells three times in PBS after incubation. If desired, probe with specific primary antibodies followed by secondary antibodies conjugated to any compatible fluorophores before using DyLight 550-Phalloidin conjugate.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phalloidin
300 units/mL
DY550-Phalloidin, F-actin toxin, Amanita phalloides
300 U
Isotype Control (Primary)
-20°C
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
DyLight 550
purified
RUO
Multi-species
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.