missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PDGF-B Recombinant Protein
Description
This product contains purified recombinant human PDGF B protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
This gene encodes a member of the protein family comprised of both platelet-derived growth factors (PDGF) and vascular endothelial growth factors (VEGF). The encoded preproprotein is proteolytically processed to generate platelet-derived growth factor subunit B, which can homodimerize, or alternatively, heterodimerize with the related platelet-derived growth factor subunit A. These proteins bind and activate PDGF receptor tyrosine kinases, which play a role in a wide range of developmental processes. Mutations in this gene are associated with meningioma. Reciprocal translocations between chromosomes 22 and 17, at sites where this gene and that for collagen type 1, alpha 1 are located, are associated with dermatofibrosarcoma protuberans, a rare skin tumor. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Accession Number | P01127 |
| Concentration | 0.11 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 0.025 M sodium acetate with 1 mM EDTA, 50% glycerol and no preservative; pH 4.8 |
| Gene ID (Entrez) | 5155 |
| Name | Human PDGF-B |
| pH Range | 4.8 |
| Quantity | 10 μg |
| Source | E. coli |
| Immunogen | N/A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?