missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PDGF-A Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP75690
Code : LD
Additional Details : Weight : 0.01000kg
Description
PDGF A (human) Recombinant Protein, RP-75690, from Invitrogen.
RP-75690 contains purified recombinant human PDGF A protein with an N-terminal hexahistidine tag. RP-75690 may be used as a positive control in many immunological assays.Specifications
P04085 | |
25mM sodium acetate with 1mM EDTA, 50% glycerol and no preservative | |
Human PDGF-A | |
E. coli | |
-20°C, Avoid Freeze/Thaw Cycles | |
Human PDGF-A | |
Human | |
Hu | |
Recombinant | |
Recombinant |
Control, Western Blot | |
5154 | |
1mg | |
N/A | |
RUO | |
PDGFA | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
in vitro research only.