missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PTH Recombinant Protein
Description
This is a recombinant protein suitable for use as a control in ELISA and Western blot applications. Reconstitute the lyophilized product in sterile water not less than 100 μg/mL.
This gene encodes a member of the parathyroid family of proteins. The encoded preproprotein is proteolytically processed to generate a protein that binds to the parathyroid hormone/parathyroid hormone-related peptide receptor and regulates blood calcium and phosphate levels. Excess production of the encoded protein, known as hyperparathyroidism, can result in hypercalcemia and kidney stones. On the other hand, defective processing of the encoded protein may lead to hypoparathyroidism, which can result in hypocalcemia and numbness.
Specifications
Specifications
| Accession Number | P01270 |
| For Use With (Application) | Control |
| Formulation | 1.15 mg sodium citrate, citric acid with 0.1117 mg EDTA, 0.2 mg Tween 80, 50 mg mannitol, 7.31 mg NaCl and no preservative |
| Gene ID (Entrez) | 5741 |
| Molecular Weight (g/mol) | 4.1 kDa |
| Name | Human PTH |
| Quantity | 100 μg |
| Source | E. coli |
| Immunogen | N/A |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?