missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PON2 Recombinant Protein
Description
This product contains purified recombinant human Paraoxonase 2 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
This gene encodes a member of the paraoxonase gene family, which includes three known members located adjacent to each other on the long arm of chromosome 7. The encoded protein is ubiquitously expressed in human tissues, membrane-bound, and may act as a cellular antioxidant, protecting cells from oxidative stress. Hydrolytic activity against acylhomoserine lactones, important bacterial quorum-sensing mediators, suggests the encoded protein may also play a role in defense responses to pathogenic bacteria. Mutations in this gene may be associated with vascular disease and a number of quantitative phenotypes related to diabetes. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
Specifications
| Accession Number | Q15165 |
| Concentration | 0.10 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 20 mM tris HCl with 50% glycerol and no preservative; pH 8 |
| Gene ID (Entrez) | 5445 |
| Name | Human PON2 |
| pH Range | 8 |
| Quantity | 10 μg |
| Source | E. coli |
| Immunogen | N/A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?