missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BACE2 (aa 441-457) Synthetic Peptide

Product Code. 11564820
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11564820 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11564820 Supplier Invitrogen™ Supplier No. PEP166

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

Human BACE2 (residues 441-457) Synthetic Peptide, PEP-166, from Invitrogen.

BACE2 or Beta Secretase 2 is responsible for the proteolytic processing of the amyloid precursor protein (APP). BACE2 cleaves APP between residues 690 and 691. This cleavage leads to the generation and extracellular release of beta-cleaved soluble APP. BACE2 is also know to cleave APP between residues 671 and 672. Accumulation of APP is a signature of Alzheimer disease and frequent complication of adult Down syndrome patients. PEP-166 is an 18 amino acid synthetic peptide whose sequence is derived from human BACE2 protein. The sequence of this peptide is (amino to carboxy terminus): V (441)-S-E-I-S-G-P-F-S-T-E-D-I-A-S-N-C (457)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human BACE2, catalog &Num; PA1-753. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 mL of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 25825
Species Human
Purification Method purified
Accession Number Q9Y5Z0
Common Name Human BACE2 (aa 441-457)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol BACE2
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence V(441)-S-E-I-S-G-P-F-S-T-E-D-I-A-S-N-C(457)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.