missing translation for 'onlineSavingsMsg'
Learn More

Human alpha-1a Adrenergic Receptor (aa 339-349), Synthetic Peptide, Invitrogen™

Product Code. 11574860
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 11574860

Brand: Invitrogen PEP216

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Synthetic Peptide

Alpha 1 Adrenergic Receptor (residues 339-349) Synthetic Peptide, PEP-216, from Invitrogen.

PEP-216 is a 11 amino acid synthetic peptide. The sequence of this peptide is (amino to carboxy terminus: K (339) F S R E K K A A K T (349)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA1-047. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 148
Species Human
Purification Method purified
Accession Number P35348
Common Name Human alpha-1a Adrenergic Receptor (aa 339-349)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol ADRA1A
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence K(339) F S R E K K A A K T(349)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.