missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FABP8 (M-FABP) Recombinant Protein
Description
This product contains purified recombinant human myelin FABP protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
The protein encoded by this gene localizes to myelin sheaths of the peripheral nervous system. The encoded protein can bind both the membrane layers of the sheaths and monomeric lipids, and is thought to provide stability to the sheath. A defect in this gene was shown to be a cause of dominant demyelinating CMT neuropathy.
Specifications
Specifications
| Accession Number | P02689 |
| Concentration | 0.22 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 20 mM tris HCl with 50% glycerol and no preservative; pH 8 |
| Gene ID (Entrez) | 5375 |
| Name | Human FABP8 (M-FABP) |
| Quantity | 10 μg |
| Source | E. coli |
| Immunogen | N/A |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?