missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CXCL10 (IP-10) Recombinant Protein

Product Code. 11528971
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11528971 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11528971 Supplier Invitrogen™ Supplier No. RP75646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This product contains purified recombinant human IFN Inducible Protein protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02778
Concentration 0.3 mg/mL
For Use With (Application) Control, Western Blot
Formulation 20 mM tris HCl with 50% glycerol and no preservative; pH 8
Gene ID (Entrez) 3627
Name Human CXCL10 (IP-10)
Quantity 1 mg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias 10 kDa interferon gamma-induced protein; 10 kDa interferon-gamma induced protein precursor; C Cmotif chemokine; C x C motif chemokine; C7; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 10; chemokine C-X-C motif ligand 10; chemokine CXC-like protein; Crg2; CRG-2; CXC; CXC chemokine; CXC motif chemokine; C-X-C motif chemokine 10; C-X-C motif chemokine 10-like protein; C-X-C motif chemokine ligand 10; CXCL; CXCL10; CXCL10(1-73); gamma interferon inducible protein 10; gamma IP10; gamma-IP10; gIP-10; H-IP-10; IFI10; Inp10; interferon activated gene 10; interferon-gamma induced protein CRG-2; interferon-gamma-induced protein CRG-2; interferon-gamma-inducible protein 10; interferon-gamma-inducible protein-10; interferon-inducible cytokine IP-10; interferon-inducible protein 10; IP10; IP-10; M-IP-10; Mob1; mob-1; protein 10 from interferon (gamma)-induced cell line; protein Mob-1; Scyb10; similar to Small inducible cytokine B10 precursor (CXCL10) (10 kDa interferon-gamma induced protein) (Gamma-IP10) (IP-10); small inducible cytokine B subfamily (Cys-X-Cys) member 10; small inducible cytokine B subfamily (Cys-X-Cys), member 10; small inducible cytokine subfamily B (Cys-X-Cys), member 10; small-inducible cytokine B10
Common Name CXCL10 (IP-10)
Gene Symbol CXCL10
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Val22-Pro98 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.