missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN gamma, Recombinant Protein, Invitrogen™
Description
Interferon gamma (human) Recombinant Protein, RP-75630, from Invitrogen.
RP-75630 contains purified recombinant human IFN gamma protein with an N-terminal hexahistidine tag. RP-75630 may be used as a positive control in many immunological assays.
Specifications
Specifications
| Accession Number | P01579 |
| Concentration | 0.1mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 20mM tris HCl with 50% glycerol and no preservative; pH 8 |
| Gene ID (Entrez) | 3458 |
| Name | Human IFN gamma |
| pH Range | 8 |
| Quantity | 100 μg |
| Source | E. coli |
| Immunogen | N/A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?