missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ILK Recombinant Protein
Description
This product contains a purified 452 amino acid fragment (aa1-452) of mature human ILK1. This product may be used as a positive control in many immunological assays.
Transduction of extracellular matrix signals through integrins influences intracellular and extracellular functions, and appears to require interaction of integrin cytoplasmic domains with cellular proteins. Integrin-linked kinase (ILK), interacts with the cytoplasmic domain of beta-1 integrin. This gene encodes a serine/threonine protein kinase with 4 ankyrin-like repeats, which associates with the cytoplasmic domain of beta integrins and acts as a proximal receptor kinase regulating integrin-mediated signal transduction. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
| Accession Number | Q13418 |
| Concentration | 0.113 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 25 mM sodium acetate with 50% glycerol and no preservative; pH 4.8 |
| Gene ID (Entrez) | 3611 |
| Molecular Weight (g/mol) | 55.92 kDa |
| Name | Human ILK |
| Purification Method | SDS-PAGE |
| Quantity | 100 μg |
| Source | E. coli |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?