missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human INHA Recombinant Protein

Product Code. 11588951
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11588951 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11588951 Supplier Invitrogen™ Supplier No. RP75632

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

RP-75632 contains a purified 134 amino acid fragment (aa233-366) corresponding to the mature Inhibin alpha subunit protein with an N-terminal hexahistidine tag. RP-75632 may be used as a positive control in Western blot assays.

The inhibin alpha subunit joins either the beta A or beta B subunit to form a pituitary FSH secretion inhibitor. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumor-suppressor activity. In addition, serum levels of inhibin have been shown to reflect the size of granulosa-cell tumors and can therefore be used as a marker for primary as well as recurrent disease. However, in prostate cancer, expression of the inhibin alpha-subunit gene was suppressed and was not detectable in poorly differentiated tumor cells. Furthermore, because expression in gonadal and various extragonadal tissues may vary severalfold in a tissue-specific fashion, it is proposed that inhibin may be both a growth/differentiation factor and a hormone.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05111
Concentration 0.05 mg/mL
For Use With (Application) Control, Western Blot
Formulation Laemmli with no preservative; pH 6.8
Gene ID (Entrez) 3623
Molecular Weight (g/mol) 19.2 kDa
Name Human INHA
pH Range 6.8
Quantity 10 μg
Source E. coli
Regulatory Status RUO
Gene Alias A inhibin subunit; A-inhibin subunit; AW555078; IHA; Inha; inhibin; inhibin alpha; inhibin alpha chain; inhibin alpha subunit
Common Name INHA
Gene Symbol INHA
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Ser233- Ile366 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.