missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-1RA Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP75580
Code : LD
Additional Details : Weight : 0.01000kg
Description
IL-1 Receptor Antagonist (human) Recombinant Protein, RP-75580, from Invitrogen.
RP-75580 contains purified recombinant human IL-1 Receptor Antagonist protein with an N-terminal hexahistidine tag. RP-75580 may be used as a positive control in many immunological assays.Specifications
P18510 | |
tris HCl with 50% glycerol and no preservative | |
Human IL1RA | |
E. coli | |
-20°C, Avoid Freeze/Thaw Cycles | |
Human IL1RA | |
Human | |
Hu | |
Recombinant | |
Recombinant |
Control, Western Blot | |
3557 | |
1mg | |
N/A | |
RUO | |
IL1RN | |
Unconjugated | |
E. coli | |
Liquid | |
purified |