missing translation for 'onlineSavingsMsg'
Learn More

Human IGF1, Recombinant Protein, Invitrogen™

Product Code. 11504930
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11504930 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11504930 Supplier Invitrogen Supplier No. RP10910

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

IGF1 Recombinant Protein, RP-10910, from Invitrogen.

The RP-10910 protein is a recombinant protein suitable for use as a control in various applications. Purity: Greater than 98% by RP-HPLC, anion-exchange chromatography; reducing and non-reducing SDS-PAGE, silver stain. Dimers/Aggregates: <1% by SDSPAGE Biological Activity: Fully biologically active compared to reference standards. Specific activity is 1 x 106 IU/mg. Biological activity at 0.2-20ng/ml. Endotoxin: <0.1ng/ug Protein Content: Determined spectrophotometrically at 280nm (0.502ext. coeff), and by RP-HPLC analysis against a calibrated standard. Lyophilized product is stable at room temperature for 3 weeks. Store lyophilized product desiccated at -20°C. Upon reconstitution, store at 4°C for 2-7 days or aliquot and store at -20°C. For long-term storage add either HSA or BSA to 0.1%. Avoid repeated freeze-thaw cycles.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05019
For Use With (Application) Control
Formulation protein with no preservative
Gene ID (Entrez) 3479
Name Human IGF1
Purification Method SDS-PAGE
Quantity 100 μg
Source E. coli
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias IGF-I, IGF-1, Somatomedin C
Common Name Human IGF1
Gene Symbol IGF1
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.