missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Yeast GSR, Recombinant Protein

Product Code. 10849720
Click to view available options
Quantity:
0.5 mg
Unit Size:
100 units
This item is not returnable. View return policy

Product Code. 10849720

Brand: Invitrogen™ LFP0071

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

Glutathione Reductase (Ys) Recombinant Protein, LF-P0071, from Invitrogen.

Glutathione reductase (GR) is a member of pyridine nucleotide-disulfide oxidoreductases, which includes the closely related enzymes thioredoxin reductase, lipoamide dehydrogenase, trypanothione reductase and mercuric ion reductase. GR is a cytoplasmic flavoenzyme widely distributed in aerobic organisms. The dimeric protein is composed of two identical subunits, each containing 1 FAD and 1 redox-active disulfide/dithiol as components of the catalytic apparatus. It plays a role in maintaining glutathione (GSH) in its reduced form by catalyzing the reduction of glutathione disulfide (GSSG). In most eukaryotic cells, GR maintains the ratio of [GSH]/[GSSG] elevated, and participates in several vital functions such as the detoxification of reactive oxygen species as well as protein and DNA biosynthesis. This protein corresponds to yeast Glutathione Reductase (1-483 aa), which has a molecular weight of 53 kDa. Specific activity: 85 U/mg, where one unit will reduce 1 umol of oxidized glutathione per min at pH 7.5 at 25°C. Purity: >95% by SDS-PAGE.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
For Use With (Application) Control
Formulation 0.02M HEPES with 10% glycerol and no preservative; pH 7.0
Name Yeast GSR
pH Range 7.0
Quantity 0.5 mg
Source E. coli
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Common Name Yeast GSR
Specific Reactivity Yeast
Conjugate Unconjugated
Cross Reactivity Ys
Species E. coli
Recombinant Recombinant
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitro research only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.