missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse Flt-3 Ligand (FLT3L) Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP8665
Code : LE
Additional Details : Weight : 0.01000kg
Description
FLT3 Ligand (mouse) Recombinant Protein, RP-8665, from Invitrogen.
FMS-related Tyrosine Kinase 3 Ligand (FLT-3 Ligand) is a growth factor important for the proliferation of hematopoietic cells. FLT-3 Ligand binds to, and transmits signals through, the receptor tyrosine kinase known as FMS-like Tyrosine Kinase-3 (FLT-3). FLT-3 Ligand promotes long-term expansion and differentiation of human pro-B cells in the presence of IL-7 or in combination of IL-7 and IL-3. Human FLT-3 Ligand can stimulate the proliferation of cells expressing murine FLT-3 receptors. Recombinant mouse IL-15 is a non-glycosylated protein, containing 163 amino acids, with a molecular weight of 18.6 kDa. Purity is typically greater than 98% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the dose-dependent induction of CTLL-2 cell proliferation and is typically less than 5 ng/mL. The specific activity of this protein is 2x10^5 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: MTPDCYFSHS PISSNFKVKF RELTDHLLKD YPVTVAVNLQ DEKHCKALWS LFLAQRWIEQ LKTVAGSKMQ TLLEDVNTEI HFVTSCTFQP LPECLRFVQT NISHLLKDTC TQLLALKPCI GKACQNFSRC LEVQCQPDSS TLLPPRSPIA LEATELPEPR PRQ.Specifications
P49772 | |
Control | |
14256 | |
Mouse Flt3 Ligand (Flt3L) | |
E. coli | |
RUO | |
Mouse Flt3 Ligand (Flt3L) | |
Mouse | |
Ms | |
Recombinant | |
Recombinant |
0.1mg/mL | |
10mM sodium phosphate with no preservative | |
∽16.4 | |
10μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
M-FLT-3-L | |
Flt3l | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |
in vitroresearch only