missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ pH Combination Electrode

Product Code. 11778146
Change view
Click to view available options
Connector Types:
6-pin
BNC
Material:
Epoxy
Stainless Steel
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Material unitSize
11778146 6-pin Stainless Steel Each
12983407 BNC Epoxy Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11778146 Supplier Thermo Scientific™ Supplier No. 01X234601

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Perform accurate pH measurements with the Thermo Scientific™ pH Combination Electrode.

Thermo Scientific™ offers an extensive assortment of quality pH electrodes and sensors. Available in a variety of body materials, body styles, fill types, reference styles, and junctions to meet the needs of your sample measurements.

Our meters, electrodes, buffers, standards, and solutions are designed for a number of applications and industries:

  • General laboratory testing
  • R&D
  • Government compliance
  • Pharmaceutical
  • Power generation
  • Drinking water
  • Wastewater treatment
  • Food & beverage testing
  • Environment
  • General industrial processing
TRUSTED_SUSTAINABILITY

Specifications

Connector Types 6-pin
Material Stainless Steel
pH Range 0 to 13
Temperature (Metric) Operating 0°C to 80°C
Type Combination pH Electrode
Temperature (Metric) Range 0°C to 80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.